Product Details

SNP ID
rs75761252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67066635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCTGAGGCAAAAAACTAGGCCCT[A/G]TCCCCAGAAGCTCCCCATCTGATGA
Phenotype
MIM: 605781
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RHOD PubMed Links

Gene Details

Gene
RHOD
Gene Name
ras homolog family member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300886.1 Intron NP_001287815.1
NM_014578.3 Intron NP_055393.1

View Full Product Details