Product Details

SNP ID
rs75547926
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56375731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGCTATTCTTATCCATCTACCT[C/T]TTCACAGTGGTAGGCAACTTGGGTT
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OR8U1 PubMed Links

Gene Details

Gene
OR8U1
Gene Name
olfactory receptor family 8 subfamily U member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005204.1 108 Silent Mutation CTC,CTT L36L NP_001005204.1

View Full Product Details