Product Details

SNP ID
rs76044076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56375740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTATCCATCTACCTCTTCACAGT[A/G]GTAGGCAACTTGGGTTTGATCCTAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR8U1 PubMed Links

Gene Details

Gene
OR8U1
Gene Name
olfactory receptor family 8 subfamily U member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005204.1 117 Silent Mutation GTA,GTG V39V NP_001005204.1

View Full Product Details