Product Details

SNP ID
rs77515824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56375781 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGATCCTACTCATTAGAGCGGATA[A/C]AAGTCTCAACACACCAATGTACTTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR8U1 PubMed Links

Gene Details

Gene
OR8U1
Gene Name
olfactory receptor family 8 subfamily U member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005204.1 158 Missense Mutation AAA,ACA K53T NP_001005204.1

View Full Product Details