Product Details

SNP ID
rs74416040
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57181924 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAAGCCCCACCTTCACAGACGCT[C/G]GCCCCAGATGTAGCTGCCTTCCTGT
Phenotype
MIM: 615213
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LRRC55 PubMed Links

Gene Details

Gene
LRRC55
Gene Name
leucine rich repeat containing 55
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005210.2 178 Missense Mutation CGC,GGC R11G NP_001005210.1

View Full Product Details