Product Details
- SNP ID
-
rs77262037
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:83459914 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATATATATCTTCTAAAGTATCTCC[C/T]TGGACAATAGCTGTAACAAAAGCAA
- Phenotype
-
MIM: 603583
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DLG2
PubMed Links
Gene Details
- Gene
- DLG2
- Gene Name
- discs large MAGUK scaffold protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001142699.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q944Q |
NP_001136171.1 |
NM_001142700.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q718Q |
NP_001136172.1 |
NM_001142702.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q303Q |
NP_001136174.1 |
NM_001206769.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q878Q |
NP_001193698.1 |
NM_001300983.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q821Q |
NP_001287912.1 |
NM_001364.3 |
2350 |
Silent Mutation |
CAA,CAG |
Q839Q |
NP_001355.2 |
XM_005273810.4 |
2350 |
Silent Mutation |
CAA,CAG |
Q806Q |
XP_005273867.1 |
XM_005273811.4 |
2350 |
Silent Mutation |
CAA,CAG |
Q788Q |
XP_005273868.1 |
XM_011544778.2 |
2350 |
Silent Mutation |
CAA,CAG |
Q994Q |
XP_011543080.1 |
XM_011544780.2 |
2350 |
Silent Mutation |
CAA,CAG |
Q976Q |
XP_011543082.1 |
XM_011544782.2 |
2350 |
Silent Mutation |
CAA,CAG |
Q860Q |
XP_011543084.1 |
XM_011544788.2 |
2350 |
Silent Mutation |
CAA,CAG |
Q736Q |
XP_011543090.1 |
XM_017017254.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q1008Q |
XP_016872743.1 |
XM_017017255.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q995Q |
XP_016872744.1 |
XM_017017256.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q995Q |
XP_016872745.1 |
XM_017017257.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q994Q |
XP_016872746.1 |
XM_017017258.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q954Q |
XP_016872747.1 |
XM_017017259.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q938Q |
XP_016872748.1 |
XM_017017260.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q937Q |
XP_016872749.1 |
XM_017017261.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q924Q |
XP_016872750.1 |
XM_017017262.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q892Q |
XP_016872751.1 |
XM_017017263.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q874Q |
XP_016872752.1 |
XM_017017264.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q873Q |
XP_016872753.1 |
XM_017017265.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q872Q |
XP_016872754.1 |
XM_017017266.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q860Q |
XP_016872755.1 |
XM_017017267.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q853Q |
XP_016872756.1 |
XM_017017268.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q852Q |
XP_016872757.1 |
XM_017017269.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q849Q |
XP_016872758.1 |
XM_017017270.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q842Q |
XP_016872759.1 |
XM_017017271.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q841Q |
XP_016872760.1 |
XM_017017272.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q840Q |
XP_016872761.1 |
XM_017017273.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q840Q |
XP_016872762.1 |
XM_017017274.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q835Q |
XP_016872763.1 |
XM_017017275.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q826Q |
XP_016872764.1 |
XM_017017276.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q817Q |
XP_016872765.1 |
XM_017017277.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q811Q |
XP_016872766.1 |
XM_017017278.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q808Q |
XP_016872767.1 |
XM_017017279.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q790Q |
XP_016872768.1 |
XM_017017280.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q789Q |
XP_016872769.1 |
XM_017017281.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q789Q |
XP_016872770.1 |
XM_017017282.1 |
2350 |
Intron |
|
|
XP_016872771.1 |
XM_017017283.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q779Q |
XP_016872772.1 |
XM_017017284.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q770Q |
XP_016872773.1 |
XM_017017285.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q765Q |
XP_016872774.1 |
XM_017017286.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q726Q |
XP_016872775.1 |
XM_017017287.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q387Q |
XP_016872776.1 |
XM_017017288.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q335Q |
XP_016872777.1 |
XM_017017289.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q362Q |
XP_016872778.1 |
XM_017017290.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q335Q |
XP_016872779.1 |
XM_017017291.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q332Q |
XP_016872780.1 |
XM_017017292.1 |
2350 |
Silent Mutation |
CAA,CAG |
Q330Q |
XP_016872781.1 |
View Full Product Details