Product Details

SNP ID
rs74977878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:94025976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGCCACTAGAGACTGGAGTATA[A/G]TGTCTTTAACAAATTAGGATTTTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HEPHL1 PubMed Links

Gene Details

Gene
HEPHL1
Gene Name
hephaestin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098672.1 Intron NP_001092142.1

View Full Product Details