Product Details

SNP ID
rs116327135
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:184605914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGAGTAATAAATAATTAAAATT[C/T]AATCATGGTTGTCAAAGTAAACAAA
Phenotype
MIM: 612282
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105373780 PubMed Links

Gene Details

Gene
LOC105373780
Gene Name
proline-rich protein 18-like
There are no transcripts associated with this gene.

Gene
ZNF804A
Gene Name
zinc finger protein 804A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194250.1 Intron NP_919226.1

View Full Product Details