Product Details

SNP ID
rs116767088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:196963149 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGGAATACATAAAGATGACAGA[C/T]GCTTCATCTCAAGACAAGCTTCCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD44 PubMed Links

Gene Details

Gene
ANKRD44
Gene Name
ankyrin repeat domain 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195144.1 Intron NP_001182073.1
NM_153697.2 Intron NP_710181.2
XM_005246945.1 Intron XP_005247002.1
XM_005246946.1 Intron XP_005247003.1
XM_005246947.2 Intron XP_005247004.1
XM_005246948.2 Intron XP_005247005.1
XM_006712838.3 Intron XP_006712901.1
XM_011512148.2 Intron XP_011510450.1
XM_017005252.1 Intron XP_016860741.1

View Full Product Details