Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001042599.1 | 9523 | UTR 3 | NP_001036064.1 | ||
NM_005235.2 | 9523 | UTR 3 | NP_005226.1 | ||
XM_005246376.2 | 9523 | UTR 3 | XP_005246433.1 | ||
XM_005246377.2 | 9523 | UTR 3 | XP_005246434.1 | ||
XM_006712364.2 | 9523 | UTR 3 | XP_006712427.1 | ||
XM_017003577.1 | 9523 | UTR 3 | XP_016859066.1 | ||
XM_017003578.1 | 9523 | UTR 3 | XP_016859067.1 | ||
XM_017003579.1 | 9523 | UTR 3 | XP_016859068.1 | ||
XM_017003580.1 | 9523 | UTR 3 | XP_016859069.1 | ||
XM_017003581.1 | 9523 | UTR 3 | XP_016859070.1 | ||
XM_017003582.1 | 9523 | UTR 3 | XP_016859071.1 |