Product Details

SNP ID
rs114609904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:240044795 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAATATTTAGTACTTTAACACAC[C/T]GCAATTTATTGGGGACTTTAAAATC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR6B3 PubMed Links

Gene Details

Gene
OR6B3
Gene Name
olfactory receptor family 6 subfamily B member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173351.1 1675 Intron NP_775486.1
XM_011510671.2 1675 UTR 3 XP_011508973.1
Gene
PRR21
Gene Name
proline rich 21
There are no transcripts associated with this gene.

View Full Product Details