Product Details

SNP ID
rs114261595
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:118900823 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAGAACCATCAAATGGAAAAAAA[A/C]ATAAGTAATGCTAAAGTAAATTCAG
Phenotype
MIM: 608351
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IGSF11 PubMed Links

Gene Details

Gene
IGSF11
Gene Name
immunoglobulin superfamily member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015887.1 3240 UTR 3 NP_001015887.1
NM_152538.2 3240 UTR 3 NP_689751.2
XM_006713517.1 3240 UTR 3 XP_006713580.1
XM_011512462.2 3240 UTR 3 XP_011510764.1
XM_011512463.1 3240 UTR 3 XP_011510765.1
XM_011512464.2 3240 UTR 3 XP_011510766.1
XM_011512465.1 3240 UTR 3 XP_011510767.1
XM_011512466.2 3240 UTR 3 XP_011510768.1
XM_011512467.2 3240 UTR 3 XP_011510769.1
XM_011512468.1 3240 UTR 3 XP_011510770.1
XM_011512469.1 3240 UTR 3 XP_011510771.1
XM_011512470.1 3240 UTR 3 XP_011510772.1
XM_011512471.2 3240 UTR 3 XP_011510773.1
XM_011512472.1 3240 UTR 3 XP_011510774.1
XM_011512474.2 3240 Intron XP_011510776.1
XM_017005789.1 3240 UTR 3 XP_016861278.1
XM_017005790.1 3240 UTR 3 XP_016861279.1

View Full Product Details