Product Details

SNP ID
rs114681871
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:133040822 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCCCACTAGTGTTGCTTTCAAA[A/G]CAGCAACAAAATCATTTCCTTTCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM108 PubMed Links

Gene Details

Gene
TMEM108
Gene Name
transmembrane protein 108
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136469.2 Intron NP_001129941.1
NM_001282865.1 Intron NP_001269794.1
NM_023943.3 Intron NP_076432.1
XM_011513097.2 Intron XP_011511399.1
XM_011513098.2 Intron XP_011511400.1
XM_017007080.1 Intron XP_016862569.1
XM_017007081.1 Intron XP_016862570.1
XM_017007082.1 Intron XP_016862571.1

View Full Product Details