Product Details
- SNP ID
-
rs114672668
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:139044261 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGGAAAACGTCCACTTCCAGGCC[A/G]GCAGACCAGTCGCCCTGCGCTCCTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRR23C
PubMed Links
Gene Details
- Gene
- PRR23C
- Gene Name
- proline rich 23C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001134657.1 |
632 |
Silent Mutation |
GCC,GCT |
A120A |
NP_001128129.1 |
View Full Product Details