Product Details

SNP ID
rs115386662
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:100496470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAACCAGTCTAACTTTGTAGCCC[A/G]AGCCTTACCCAATAGGCTATGCTAC
Phenotype
MIM: 616928
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM45A PubMed Links

Gene Details

Gene
TMEM45A
Gene Name
transmembrane protein 45A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018004.2 Intron NP_060474.1
XM_005247569.2 Intron XP_005247626.1
XM_017006715.1 Intron XP_016862204.1

View Full Product Details