Product Details

SNP ID
rs115136515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:180918006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATATATTTCTTGTAAAGTTTTG[G/T]GTTTTGTATTAGAAGAGCAACACAT
Phenotype
MIM: 600819
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FXR1 PubMed Links

Gene Details

Gene
FXR1
Gene Name
FMR1 autosomal homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013438.2 Intron NP_001013456.1
NM_001013439.2 Intron NP_001013457.1
NM_005087.3 Intron NP_005078.2
XM_005247813.3 Intron XP_005247870.2
XM_005247814.3 Intron XP_005247871.2
XM_005247815.3 Intron XP_005247872.2
XM_005247816.3 Intron XP_005247873.2
XM_006713775.3 Intron XP_006713838.1
XM_017007298.1 Intron XP_016862787.1
XM_017007299.1 Intron XP_016862788.1
XM_017007300.1 Intron XP_016862789.1
XM_017007301.1 Intron XP_016862790.1

View Full Product Details