Product Details

SNP ID
rs113994046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184136636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCCTCTTGCCCCTGGCCAATGTG[A/G]CATTAATTGACTACACTCTGGAATT
Phenotype
MIM: 603945
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF2B5 PubMed Links

Gene Details

Gene
EIF2B5
Gene Name
eukaryotic translation initiation factor 2B subunit epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003907.2 584 Missense Mutation ACA,GCA T74A NP_003898.2
XM_011513265.1 584 Intron XP_011511567.1
XM_011513266.2 584 Intron XP_011511568.1

View Full Product Details