Product Details

SNP ID
rs114149299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:41941863 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAACAGATAGGGCATTTCACCTC[C/T]TATCTTTTCTAAATACATTGTATTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM33 PubMed Links

Gene Details

Gene
TMEM33
Gene Name
transmembrane protein 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018126.2 Intron NP_060596.2
XM_005248116.3 Intron XP_005248173.1
XM_005248117.2 Intron XP_005248174.1

View Full Product Details