Product Details

SNP ID
rs114639834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:10279451 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACCCGTTTATAGTTCATGTTCTT[C/T]ATGGCTTTGCAGCATTTGTCACTTT
Phenotype
MIM: 613379
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CMBL PubMed Links

Gene Details

Gene
CMBL
Gene Name
carboxymethylenebutenolidase homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138809.3 2191 UTR 3 NP_620164.1
XM_017009034.1 2191 UTR 3 XP_016864523.1
XM_017009035.1 2191 UTR 3 XP_016864524.1

View Full Product Details