Product Details

SNP ID
rs113860223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:39284359 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCTAATGAATAAGATTGTACAA[C/T]CAATGTTAATCAGCTTCTCAGGTAG
Phenotype
MIM: 120940 MIM: 602731
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C9 PubMed Links

Gene Details

Gene
C9
Gene Name
complement component 9
There are no transcripts associated with this gene.

Gene
FYB
Gene Name
FYN binding protein
There are no transcripts associated with this gene.

View Full Product Details