Product Details

SNP ID
rs116939630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:60754695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCTCTGACCTTTGGTGTAAGCAC[A/G]GTACCAAAAATGGAGAAAGAGCAGC
Phenotype
MIM: 614451
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ELOVL7 PubMed Links

Gene Details

Gene
ELOVL7
Gene Name
ELOVL fatty acid elongase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104558.1 1342 Missense Mutation CGT,TGT R259C NP_001098028.1
NM_001297617.1 1342 Missense Mutation CGT,TGT R246C NP_001284546.1
NM_001297618.1 1342 Missense Mutation CGT,TGT R172C NP_001284547.1
NM_024930.2 1342 Missense Mutation CGT,TGT R259C NP_079206.2
XM_005248606.4 1342 Missense Mutation CGT,TGT R259C XP_005248663.1
XM_005248607.3 1342 Missense Mutation CGT,TGT R246C XP_005248664.1
XM_006714695.3 1342 Missense Mutation CGT,TGT R259C XP_006714758.1
XM_011543651.2 1342 Missense Mutation CGT,TGT R259C XP_011541953.1
XM_017009885.1 1342 Missense Mutation CGT,TGT R259C XP_016865374.1
XM_017009886.1 1342 Missense Mutation CGT,TGT R246C XP_016865375.1
XM_017009887.1 1342 Missense Mutation CGT,TGT R246C XP_016865376.1
XM_017009888.1 1342 Intron XP_016865377.1
XM_017009889.1 1342 Intron XP_016865378.1

View Full Product Details