Product Details

SNP ID
rs115682782
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:76407474 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTGAATAAGTCGGTATTTAGCAT[A/G]GCTCAGCTGTAACATCCTATCAGTG
Phenotype
MIM: 605401
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IQGAP2 PubMed Links

Gene Details

Gene
IQGAP2
Gene Name
IQ motif containing GTPase activating protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285460.1 Intron NP_001272389.1
NM_001285461.1 Intron NP_001272390.1
NM_001285462.1 Intron NP_001272391.1
NM_006633.3 Intron NP_006624.2
XM_005248410.2 Intron XP_005248467.1
XM_005248414.1 Intron XP_005248471.1
XM_011543108.1 Intron XP_011541410.1
XM_017008960.1 Intron XP_016864449.1

View Full Product Details