Product Details

SNP ID
rs111926973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:75756046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTACCAACCTGCTTTGATAAAT[A/G]TAAGTATGTTCCATCATTTATCACA
Phenotype
MIM: 600970
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYO6 PubMed Links

Gene Details

Gene
MYO6
Gene Name
myosin VI
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300899.1 Intron NP_001287828.1
NM_004999.3 Intron NP_004990.3
XM_005248719.3 Intron XP_005248776.1
XM_005248720.3 Intron XP_005248777.1
XM_005248721.3 Intron XP_005248778.1
XM_005248722.3 Intron XP_005248779.1
XM_005248724.3 Intron XP_005248781.1
XM_005248726.3 Intron XP_005248783.1
XM_017010899.1 Intron XP_016866388.1

View Full Product Details