Product Details

SNP ID
rs116501140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:77461894 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTTCTAGTGGGCATTATCCACG[A/G]TACCAAAGCAGGGCAGGGATTCCCT
Phenotype
MIM: 182131
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HTR1B PubMed Links

Gene Details

Gene
HTR1B
Gene Name
5-hydroxytryptamine receptor 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000863.2 2129 UTR 3 NP_000854.1

View Full Product Details