Product Details

SNP ID
rs114108814
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28283886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGAAAGTGAAGGAGGAAGATCCC[A/G]CCTGGGAGCAGGTGTGCAACTCACA
Phenotype
MIM: 616474
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PGBD1 PubMed Links

Gene Details

Gene
PGBD1
Gene Name
piggyBac transposable element derived 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184743.1 493 Missense Mutation ACC,GCC T25A NP_001171672.1
NM_032507.3 493 Missense Mutation ACC,GCC T25A NP_115896.1
XM_006715228.2 493 UTR 5 XP_006715291.1
XM_011514942.2 493 UTR 5 XP_011513244.1
XM_011514943.2 493 UTR 5 XP_011513245.1
XM_011514944.2 493 UTR 5 XP_011513246.1
XM_017011361.1 493 Missense Mutation ACC,GCC T25A XP_016866850.1
XM_017011362.1 493 UTR 5 XP_016866851.1
XM_017011363.1 493 UTR 5 XP_016866852.1
Gene
ZSCAN26
Gene Name
zinc finger and SCAN domain containing 26
There are no transcripts associated with this gene.

View Full Product Details