Product Details

SNP ID
rs114618932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31859728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCAGTGGCACAGCTCAGAGTGT[C/T]CCATAGACACTGATTTTGGCCACGG
Phenotype
MIM: 608272 MIM: 606107
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NEU1 PubMed Links

Gene Details

Gene
NEU1
Gene Name
neuraminidase 1 (lysosomal sialidase)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000434.3 1395 Silent Mutation GGA,GGG G413G NP_000425.1
Gene
SLC44A4
Gene Name
solute carrier family 44 member 4
There are no transcripts associated with this gene.

View Full Product Details