Product Details

SNP ID
rs114529664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35575004 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGAATATTATTCCCTAAAAGGAA[C/T]AGAGGAGGGGAGAAAAAGCACAATT
Phenotype
MIM: 602623
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FKBP5 PubMed Links

Gene Details

Gene
FKBP5
Gene Name
FK506 binding protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145775.2 2363 UTR 3 NP_001139247.1
NM_001145776.1 2363 UTR 3 NP_001139248.1
NM_001145777.1 2363 Intron NP_001139249.1
NM_004117.3 2363 UTR 3 NP_004108.1

View Full Product Details