Product Details

SNP ID
rs116447831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41789090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAATGAACTCCAAATTCCCACG[C/T]GGGTCTTGAGCAACCTGCCGTCACA
Phenotype
MIM: 607744 MIM: 611389 MIM: 616168
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FRS3 PubMed Links

Gene Details

Gene
FRS3
Gene Name
fibroblast growth factor receptor substrate 3
There are no transcripts associated with this gene.

Gene
PRICKLE4
Gene Name
prickle planar cell polarity protein 4
There are no transcripts associated with this gene.

Gene
TOMM6
Gene Name
translocase of outer mitochondrial membrane 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134493.1 Intron NP_001127965.1
Gene
USP49
Gene Name
ubiquitin specific peptidase 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286554.1 Intron NP_001273483.1
NM_018561.4 Intron NP_061031.2

View Full Product Details