Product Details

SNP ID
rs116442182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:44131894 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATCAGACTAGAGAAACTGGGAT[A/G]TAGCTGGTGCAGGCACCTCGTGGAG
Phenotype
MIM: 611827
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL14 PubMed Links

Gene Details

Gene
MRPL14
Gene Name
mitochondrial ribosomal protein L14
There are no transcripts associated with this gene.

Gene
TMEM63B
Gene Name
transmembrane protein 63B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318792.1 Intron NP_001305721.1
NM_018426.2 Intron NP_060896.1
XM_005249211.3 Intron XP_005249268.1
XM_005249213.4 Intron XP_005249270.1
XM_005249214.4 Intron XP_005249271.1
XM_005249215.4 Intron XP_005249272.1
XM_005249217.1 Intron XP_005249274.1
XM_006715135.1 Intron XP_006715198.1
XM_017010999.1 Intron XP_016866488.1
XM_017011000.1 Intron XP_016866489.1
XM_017011001.1 Intron XP_016866490.1
XM_017011002.1 Intron XP_016866491.1

View Full Product Details