Product Details

SNP ID
rs111379918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:46161001 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCCATCAGGAGAGAAATTTAAAG[A/T]GCAATACATAAGGTACTTTACATAG
Phenotype
MIM: 617001
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ENPP5 PubMed Links

Gene Details

Gene
ENPP5
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290072.1 1914 UTR 3 NP_001277001.1
NM_001290073.1 1914 UTR 3 NP_001277002.1
NM_021572.5 1914 UTR 3 NP_067547.1
XM_005249260.3 1914 UTR 3 XP_005249317.1
XM_011514786.2 1914 UTR 3 XP_011513088.1

View Full Product Details