Product Details

SNP ID
rs115865539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:24698954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATTTCATTGGTCAACTTTTAAC[A/G]TGCATATGACCTTTAACAGTTCTGA
Phenotype
MIM: 608798 MIM: 606959
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DFNA5 PubMed Links

Gene Details

Gene
DFNA5
Gene Name
DFNA5, deafness associated tumor suppressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127453.1 1896 UTR 3 NP_001120925.1
NM_001127454.1 1896 UTR 3 NP_001120926.1
NM_004403.2 1896 UTR 3 NP_004394.1
XM_017011802.1 1896 UTR 3 XP_016867291.1
Gene
MPP6
Gene Name
membrane palmitoylated protein 6
There are no transcripts associated with this gene.

View Full Product Details