Product Details

SNP ID
rs112203306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:2519880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCACCATGCTCAAGCGCTGCGGC[C/G]GGCGCCTGCTGCTGGCGCTGGCGGG
Phenotype
MIM: 602576
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LFNG PubMed Links

Gene Details

Gene
LFNG
Gene Name
LFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040167.1 36 Missense Mutation CGG,GGG R7G NP_001035257.1
NM_001040168.1 36 Missense Mutation CGG,GGG R7G NP_001035258.1
NM_001166355.1 36 Intron NP_001159827.1
NM_002304.2 36 Intron NP_002295.1
Gene
MIR4648
Gene Name
microRNA 4648
There are no transcripts associated with this gene.

View Full Product Details