Product Details
- SNP ID
-
rs115139383
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:26534841 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTCCCTTCCTCACCCCTGGGCTC[C/G]CAGGGCTCTCCCTCCCAGCCCTTCC
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA0087
PubMed Links
Gene Details
- Gene
- KIAA0087
- Gene Name
- KIAA0087
There are no transcripts associated with this gene.
View Full Product Details