Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_198455.2 | 230 | Missense Mutation | TAC,TGC | Y77C | NP_940857.2 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
XM_005249959.4 | 230 | Intron | XP_005250016.1 | ||
XM_005249960.4 | 230 | Intron | XP_005250017.1 | ||
XM_005249961.4 | 230 | Intron | XP_005250018.1 | ||
XM_006715864.3 | 230 | Intron | XP_006715927.1 | ||
XM_011515855.2 | 230 | Intron | XP_011514157.1 | ||
XM_011515856.2 | 230 | Intron | XP_011514158.1 | ||
XM_011515857.2 | 230 | Intron | XP_011514159.1 | ||
XM_011515858.1 | 230 | Intron | XP_011514160.1 | ||
XM_017011799.1 | 230 | Intron | XP_016867288.1 |