Product Details

SNP ID
rs113702561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151054225 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTCACCTACCTGCAGCAGATCC[C/T]TCCCTGTGGCATTGAGTTTGGGCAC
Phenotype
MIM: 605464 MIM: 611741 MIM: 123831 MIM: 109280
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCB8 PubMed Links

Gene Details

Gene
ABCB8
Gene Name
ATP binding cassette subfamily B member 8
There are no transcripts associated with this gene.

Gene
ASIC3
Gene Name
acid sensing ion channel subunit 3
There are no transcripts associated with this gene.

Gene
CDK5
Gene Name
cyclin dependent kinase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164410.2 800 Missense Mutation AAG,AGG K228R NP_001157882.1
NM_004935.3 800 Missense Mutation AAG,AGG K260R NP_004926.1
Gene
SLC4A2
Gene Name
solute carrier family 4 member 2
There are no transcripts associated with this gene.

View Full Product Details