Product Details

SNP ID
rs115433982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151240907 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGAGAAGTCAGTGAACTAATACA[C/T]ATAAAGTGCTTGGGACAGTGCCTGG
Phenotype
MIM: 608037 MIM: 615245 MIM: 601737
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHPF2 PubMed Links

Gene Details

Gene
CHPF2
Gene Name
chondroitin polymerizing factor 2
There are no transcripts associated with this gene.

Gene
MIR671
Gene Name
microRNA 671
There are no transcripts associated with this gene.

Gene
SMARCD3
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003801.1 Intron NP_001003801.1
NM_001003802.1 Intron NP_001003802.1
NM_003078.3 Intron NP_003069.2
XM_011516521.2 Intron XP_011514823.1
XM_017012556.1 Intron XP_016868045.1

View Full Product Details