Product Details
- SNP ID
-
rs117374561
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:143393089 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCGCAGCCATCCTGCACTATCTG[C/T]AACCTACCAGCCCTCAGCTCCCCAC
- Phenotype
-
MIM: 179610
MIM: 602002
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EPHA1
PubMed Links
Gene Details
- Gene
- EPHA1
- Gene Name
- EPH receptor A1
- Gene
- ZYX
- Gene Name
- zyxin
There are no transcripts associated with this gene.
View Full Product Details