Product Details
- SNP ID
-
rs112653404
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:87195556 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGAGTCTTTTCAACATGGTCTTCA[A/G]CATGGTCTAGAGCTTACCAGTGATC
- Phenotype
-
MIM: 608491
MIM: 616993
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DMTF1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs148183534] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DMTF1
- Gene Name
- cyclin D binding myb like transcription factor 1
- Gene
- TMEM243
- Gene Name
- transmembrane protein 243
View Full Product Details