Product Details

SNP ID
rs117656893
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100430729 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCATCCCCAAAGATATTACTGA[C/T]CCGCTCAGTCTCAATACTTGCACTG
Phenotype
MIM: 611478
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MEPCE PubMed Links

Gene Details

Gene
MEPCE
Gene Name
methylphosphate capping enzyme
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001194990.1 366 UTR 5 NP_001181919.1
NM_001194991.1 366 Intron NP_001181920.1
NM_001194992.1 366 Intron NP_001181921.1
NM_019606.5 366 Silent Mutation GAC,GAT D237D NP_062552.2
XM_011516410.2 366 Intron XP_011514712.1
Gene
PPP1R35
Gene Name
protein phosphatase 1 regulatory subunit 35
There are no transcripts associated with this gene.

Gene
ZCWPW1
Gene Name
zinc finger CW-type and PWWP domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258008.1 366 Intron NP_001244937.1
NM_017984.4 366 Intron NP_060454.3

View Full Product Details