Product Details

SNP ID
rs112132756
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101087630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCAGCCTGTGCCCTGTGTCCCCT[G/T]GTGGGTGGCACCAGCACCGGGGGGC
Phenotype
MIM: 616996
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TRIM56 PubMed Links

Gene Details

Gene
TRIM56
Gene Name
tripartite motif containing 56
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030961.2 615 Silent Mutation CTG,CTT L106L NP_112223.1
XM_011516589.2 615 Silent Mutation CTG,CTT L106L XP_011514891.1

View Full Product Details