Product Details

SNP ID
rs112250947
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:108152030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAGATAGATTGTTCTCGGCACTG[A/G]GAATATAGCAGCAAACAAAACAGAC
Phenotype
MIM: 601581
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NRCAM PubMed Links

Gene Details

Gene
NRCAM
Gene Name
neuronal cell adhesion molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037132.2 8591 Intron NP_001032209.1
NM_001193582.1 8591 Intron NP_001180511.1
NM_001193583.1 8591 Intron NP_001180512.1
NM_001193584.1 8591 Intron NP_001180513.1
NM_005010.4 8591 Intron NP_005001.3
XM_005250373.2 8591 Intron XP_005250430.1
XM_005250383.2 8591 Intron XP_005250440.1
XM_005250385.2 8591 Intron XP_005250442.1
XM_006716003.1 8591 Intron XP_006716066.1
XM_006716007.1 8591 Intron XP_006716070.1
XM_006716012.1 8591 Intron XP_006716075.1
XM_006716014.1 8591 Intron XP_006716077.1
XM_011516253.1 8591 Intron XP_011514555.1
XM_011516255.1 8591 Intron XP_011514557.1
XM_011516256.1 8591 Intron XP_011514558.1
XM_011516257.1 8591 Intron XP_011514559.1
XM_011516258.1 8591 Intron XP_011514560.1
XM_011516259.1 8591 Intron XP_011514561.1
XM_011516261.1 8591 Intron XP_011514563.1
XM_011516262.1 8591 Intron XP_011514564.1
XM_011516265.2 8591 Intron XP_011514567.1
XM_011516266.2 8591 UTR 3 XP_011514568.1
XM_011516267.1 8591 Intron XP_011514569.1
XM_011516268.2 8591 Intron XP_011514570.1
XM_011516269.1 8591 Intron XP_011514571.1
XM_011516270.1 8591 Intron XP_011514572.1
XM_011516271.1 8591 Intron XP_011514573.1
XM_017012236.1 8591 Intron XP_016867725.1
XM_017012237.1 8591 Intron XP_016867726.1
XM_017012238.1 8591 Intron XP_016867727.1
XM_017012239.1 8591 Intron XP_016867728.1
XM_017012240.1 8591 Intron XP_016867729.1
XM_017012241.1 8591 Intron XP_016867730.1
XM_017012242.1 8591 Intron XP_016867731.1
XM_017012243.1 8591 Intron XP_016867732.1
XM_017012244.1 8591 Intron XP_016867733.1
XM_017012245.1 8591 Intron XP_016867734.1
XM_017012246.1 8591 Intron XP_016867735.1
XM_017012247.1 8591 Intron XP_016867736.1
XM_017012248.1 8591 Intron XP_016867737.1
XM_017012249.1 8591 Intron XP_016867738.1
XM_017012250.1 8591 Intron XP_016867739.1
XM_017012251.1 8591 Intron XP_016867740.1
XM_017012252.1 8591 Intron XP_016867741.1
XM_017012253.1 8591 Intron XP_016867742.1
XM_017012254.1 8591 Intron XP_016867743.1
XM_017012255.1 8591 Intron XP_016867744.1
XM_017012256.1 8591 Intron XP_016867745.1
XM_017012257.1 8591 Intron XP_016867746.1
XM_017012258.1 8591 Intron XP_016867747.1
XM_017012259.1 8591 Intron XP_016867748.1

View Full Product Details