Product Details

SNP ID
rs116103307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:120791561 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAATAGTCTCTGGGGCTTAAATT[G/T]TATCTATATTGGCTTAAGATCTCCT
Phenotype
MIM: 613138
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TSPAN12 PubMed Links

Gene Details

Gene
TSPAN12
Gene Name
tetraspanin 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012338.3 Intron NP_036470.1
XM_005250239.2 Intron XP_005250296.1
XM_011515993.1 Intron XP_011514295.1
XM_011515994.1 Intron XP_011514296.1
XM_017011912.1 Intron XP_016867401.1
XM_017011913.1 Intron XP_016867402.1
XM_017011914.1 Intron XP_016867403.1

View Full Product Details