Product Details

SNP ID
rs117070316
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128028716 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTGTACCTTGTCCTTGGTATGG[G/T]TCTGAATTATATAAGGTTCAGAGAT
Phenotype
MIM: 610486 MIM: 602181
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LRRC4 PubMed Links

Gene Details

Gene
LRRC4
Gene Name
leucine rich repeat containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022143.4 2368 Missense Mutation NP_071426.1
XM_011516461.2 2368 Missense Mutation XP_011514763.1
XM_017012505.1 2368 Missense Mutation XP_016867994.1
Gene
SND1
Gene Name
staphylococcal nuclease and tudor domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017011987.1 2368 Intron XP_016867476.1

View Full Product Details