Product Details

SNP ID
rs115854463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:7796114 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATTCTAAAGTCTGTAAATCATAA[C/T]TAAGCTGGTGTTCTTACATGACTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM261 PubMed Links

Gene Details

Gene
TMEM261
Gene Name
transmembrane protein 261
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318058.1 Intron NP_001304987.1
NM_001318059.1 Intron NP_001304988.1
NM_033428.2 Intron NP_219500.1

View Full Product Details