Product Details

SNP ID
rs111505827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127909985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCGCTGACCATCCCATAGACCA[C/T]GATCTCCTCACACAGCTCCAGCGCG
Phenotype
MIM: 606378 MIM: 610135
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ST6GALNAC4 PubMed Links

Gene Details

Gene
ST6GALNAC4
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175039.3 1084 Missense Mutation ATG,GTG M229V NP_778204.1
NM_175040.3 1084 Missense Mutation ATG,GTG M145V NP_778205.1
XM_011518527.2 1084 Missense Mutation ATG,GTG M252V XP_011516829.1
XM_011518529.2 1084 UTR 3 XP_011516831.1
Gene
ST6GALNAC6
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 6
There are no transcripts associated with this gene.

View Full Product Details