Product Details

SNP ID
rs117484197
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:10559884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGAGAGCAAGCACTAGCAGAGC[C/T]GTGGGGACACCTGCAAGGAACAGAG
Phenotype
MIM: 605702
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYVE1 PubMed Links

Gene Details

Gene
LYVE1
Gene Name
lymphatic vessel endothelial hyaluronan receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006691.3 1000 Silent Mutation ACA,ACG T238T NP_006682.2
Gene
MRVI1-AS1
Gene Name
MRVI1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details