Product Details

SNP ID
rs113353493
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47259788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTTGTGCCTCTCTTTTGGAGCT[A/C]AGACTCTGCGGTGGAGCTGTGGAAG
Phenotype
MIM: 603584 MIM: 602423
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MADD PubMed Links

Gene Details

Gene
MADD
Gene Name
MAP kinase activating death domain
There are no transcripts associated with this gene.

Gene
NR1H3
Gene Name
nuclear receptor subfamily 1 group H member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130101.2 143 Intron NP_001123573.1
NM_001130102.2 143 Intron NP_001123574.1
NM_001251934.1 143 Intron NP_001238863.1
NM_001251935.1 143 Intron NP_001238864.1
NM_005693.3 143 Intron NP_005684.2
XM_005252705.1 143 Intron XP_005252762.1
XM_005252706.1 143 Intron XP_005252763.1
XM_005252707.4 143 Intron XP_005252764.1
XM_005252709.1 143 UTR 5 XP_005252766.1
XM_005252710.1 143 UTR 5 XP_005252767.1
XM_005252713.3 143 Intron XP_005252770.1
XM_005252715.2 143 Intron XP_005252772.1
XM_005252716.3 143 Intron XP_005252773.1
XM_005252718.3 143 Intron XP_005252775.1
XM_006718112.1 143 Intron XP_006718175.1
XM_006718113.1 143 Intron XP_006718176.1
XM_006718115.1 143 UTR 5 XP_006718178.1
XM_006718116.1 143 Intron XP_006718179.1
XM_011519805.2 143 Intron XP_011518107.1
XM_011519806.1 143 Intron XP_011518108.1
XM_011519807.1 143 Intron XP_011518109.1
XM_011519808.2 143 Intron XP_011518110.1
XM_017017056.1 143 Intron XP_016872545.1
XM_017017057.1 143 Intron XP_016872546.1
XM_017017058.1 143 Intron XP_016872547.1

View Full Product Details