Product Details

SNP ID
rs113950675
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47569774 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTGGGATCCCCACCTTGGACAA[C/T]CTCCAGAAGGGAGTCCAATTTGCTC
Phenotype
MIM: 601074 MIM: 603846 MIM: 609538
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CELF1 PubMed Links

Gene Details

Gene
CELF1
Gene Name
CUGBP, Elav-like family member 1
There are no transcripts associated with this gene.

Gene
KBTBD4
Gene Name
kelch repeat and BTB domain containing 4
There are no transcripts associated with this gene.

Gene
NDUFS3
Gene Name
NADH:ubiquinone oxidoreductase core subunit S3
There are no transcripts associated with this gene.

Gene
PTPMT1
Gene Name
protein tyrosine phosphatase, mitochondrial 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143984.1 523 Intron NP_001137456.1
NM_175732.2 523 Silent Mutation AAC,AAT N110N NP_783859.1

View Full Product Details