Product Details

SNP ID
rs117660271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:582056 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGAGAAGACGAGGGGGCGTCTGA[A/G]GAGGAAGACCTGGAAGACAGATCTG
Phenotype
MIM: 611780
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC143666 PubMed Links

Gene Details

Gene
LOC143666
Gene Name
uncharacterized LOC143666
There are no transcripts associated with this gene.

Gene
PHRF1
Gene Name
PHD and ring finger domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286581.1 357 Silent Mutation GAA,GAG E63E NP_001273510.1
NM_001286582.1 357 Silent Mutation GAA,GAG E62E NP_001273511.1
NM_001286583.1 357 Silent Mutation GAA,GAG E59E NP_001273512.1
NM_020901.3 357 Silent Mutation GAA,GAG E63E NP_065952.2
XM_005253025.4 357 Silent Mutation GAA,GAG E62E XP_005253082.1
XM_005253027.3 357 Silent Mutation GAA,GAG E60E XP_005253084.1
XM_011520236.2 357 Silent Mutation GAA,GAG E63E XP_011518538.1
XM_011520237.2 357 Intron XP_011518539.1

View Full Product Details