Product Details

SNP ID
rs112549328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5947426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTGTTTTGTCAGATCCCCCAGCT[A/G]GCAGCACTGGCTGTCCCTGCCCCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR56A3 PubMed Links

Gene Details

Gene
OR56A3
Gene Name
olfactory receptor family 56 subfamily A member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003443.2 80 Nonsense Mutation TAG,TGG *27W NP_001003443.2

View Full Product Details